Anti-PMP22 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108241
89320-092EA 394.91 USD
89320-092
Anti-PMP22 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to P2RY11 (purinergic receptor P2Y, G-protein coupled, 11)

  • Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PMP22
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR