Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TPD52L1 Mouse Monoclonal Antibody [clone: STo 3H9]

Supplier: Genetex

GTX10822
89337-148EA 483.46 USD InStock
89337-148
Anti-TPD52L1 Mouse Monoclonal Antibody [clone: STo 3H9]
Antibodies
Mouse monoclonal [STo 3H9] to Clathrin heavy chain

WB: Use at an assay dependent dilution. Predicted molecular weight: 201 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TPD52L1
Clonality: Monoclonal
Clone: STo 3H9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty