Anti-PDE8B Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108177
89320-082EA 394.91 USD
89320-082
Anti-PDE8B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CRF-RII (corticotropin releasing hormone receptor 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PDE8B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR