Anti-PTGES Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108159
89320-076EA 482.37 USD
89320-076
Anti-PTGES Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Metabotropic Glutamate Receptor 8 (glutamate receptor, metabotropic 8)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PTGES
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR