Anti-MYL9 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108137
89310-182EA 394.91 USD
89310-182
Anti-MYL9 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GPR10 (prolactin releasing hormone receptor)

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: MYL9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bird, Mammal
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR