Anti-ATG4B Rabbit Polyclonal Antibody

GTX108106
89310-158EA 176.03 USD
89310-158
Anti-ATG4B Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Schlafen 8 (proteasome 26S non-ATPase subunit 8 pseudogene)

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: ATG4B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Chicken, Pufferfish, Rat, Xenopus Laevis, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR