Anti-ARL6IP1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108104
89310-156EA 394.91 USD
89310-156
Anti-ARL6IP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Melanophilin (melanophilin)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ARL6IP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR