Anti-APH1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX108086
89320-060EA 394.79 USD
89320-060
Anti-APH1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SETDB2 (SET domain, bifurcated 2)

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:2000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: APH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR