Anti-MUC13 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108059
89320-046EA 582.76 USD
89320-046
Anti-MUC13 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to centaurin-alpha 2 (centaurin, alpha 2)

Suggested starting dilutions are as follows: WB: 1:500-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MUC13
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR