Anti-LRRC7 Mouse Monoclonal Antibody [clone: ICAM3.1]

Supplier: GeneTex
GTX10804
89328-200EA 630.11 USD
89328-200
Anti-LRRC7 Mouse Monoclonal Antibody [clone: ICAM3.1]
Antibodies
Mouse monoclonal [ICAM3.1] to ICAM3

ELISA: Use at an assay dependent dilution. IHC-Fr: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. RIA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 66 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LRRC7
Clonality: Monoclonal
Clone: ICAM3.1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR