Anti-WNT5B Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108021
89320-038EA 582.76 USD
89320-038
Anti-WNT5B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to AOS-1 (SUMO1 activating enzyme subunit 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: WNT5B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR