Anti-C1QTNF5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107998
89316-504EA 727.73 USD
89316-504
Anti-C1QTNF5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SYF2 (SYF2 homolog, RNA splicing factor (S. cerevisiae))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: C1QTNF5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR