Anti-SLC5A10 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107983
89320-018EA 582.76 USD
89320-018
Anti-SLC5A10 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to EPM2AIP1 (EPM2A (laforin) interacting protein 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SLC5A10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR