Anti-TICAM2 Rabbit Polyclonal Antibody

GTX107929
89310-026EA 176.03 USD
89310-026
Anti-TICAM2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to RPS6KA4 (ribosomal protein S6 kinase, 90kDa, polypeptide 4)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: TICAM2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR