Anti-CDC25A Rabbit Polyclonal Antibody

Supplier: Genetex

GTX107915
89319-994EA 394.91 USD
89319-994
Anti-CDC25A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GRK2 (adrenergic, beta, receptor kinase 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CDC25A
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR