Anti-MYL9 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107894
89319-982EA 642.92 USD
89319-982
Anti-MYL9 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ADCK1 (aarF domain containing kinase 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MYL9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR