Anti-West Nile Virus Matrix Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107815
89319-954EA 448.72 USD
89319-954
Anti-West Nile Virus Matrix Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Activin A Receptor type I (activin A receptor, type I)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: West Nile Virus Matrix
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR