Anti-CTRP7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107788
89319-944EA 448.72 USD
89319-944
Anti-CTRP7 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MAP4K6 (misshapen-like kinase 1 (zebrafish))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CTRP7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR