Anti-DOK3 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX107744
89319-926EA 394.79 USD
89319-926
Anti-DOK3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Docking protein 3 (docking protein 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: DOK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR