Anti-DRAM Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107737
89309-964EA 459.92 USD
89309-964
Anti-DRAM Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Foxp3 (forkhead box P3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: DRAM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR