Anti-ENO2 Goat Polyclonal Antibody

Supplier: Genetex

GTX10771
89316-006EA 357.98 USD
89316-006
Anti-ENO2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to IL9

ELISA: Use at a concentration of 0.5 ?g/ml. Neut: Use at a concentration of 4.1 ?g/ml. WB: Use at a concentration of 1 ?g/ml. Predicted molecular weight: 16 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ENO2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR