Anti-PDLIM3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107697
89316-484EA 745.91 USD
89316-484
Anti-PDLIM3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PDLIM3 (PDZ and LIM domain 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PDLIM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR