Anti-STIM2 Rabbit Polyclonal Antibody

GTX107685
89319-912EA 176.03 USD
89319-912
Anti-STIM2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Centaurin beta 1 (ArfGAP with coiled-coil, ankyrin repeat and PH domains 1)

Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: STIM2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR