Anti-STIM2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107684
89319-910EA 657.53 USD
89319-910
Anti-STIM2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SART3 (squamous cell carcinoma antigen recognized by T cells 3)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: STIM2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR