Anti-SLITRK6 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX107611
89319-886EA 394.91 USD
89319-886
Anti-SLITRK6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Selenophosphate synthetase 1 (selenophosphate synthetase 1)

  • Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SLITRK6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR