Anti-NANOS2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX107563
89319-868EA 394.91 USD
89319-868
Anti-NANOS2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Glycerol kinase 2 (glycerol kinase 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NANOS2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR