Anti-CAPN6 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX107538
89309-878EA 394.79 USD
89309-878
Anti-CAPN6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MLKL (mixed lineage kinase domain-like)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CAPN6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR