Anti-IP6K1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX107438
89309-850EA 394.91 USD
89309-850
Anti-IP6K1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to IP6K1 (inositol hexakisphosphate kinase 1)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: IP6K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR