Anti-ZBTB2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107431
89309-848EA 727.73 USD
89309-848
Anti-ZBTB2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to nm23-H4 (non-metastatic cells 4, protein expressed in)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ZBTB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR