Anti-ZBED3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107366
89319-834EA 582.76 USD
89319-834
Anti-ZBED3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MBNL3 (muscleblind-like 3 (Drosophila))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ZBED3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR