Anti-RBMS1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107358
89319-830EA 582.76 USD
89319-830
Anti-RBMS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RBMS1 (RNA binding motif, single stranded interacting protein 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RBMS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR