Anti-SCARA5 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX107271
89319-812EA 394.91 USD
89319-812
Anti-SCARA5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to C16orf62 (chromosome 16 open reading frame 62)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SCARA5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR