Anti-LIAR Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107270
89319-810EA 582.76 USD
89319-810
Anti-LIAR Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Drebrin (drebrin 1)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1-100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LIAR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR