Anti-NOTUM Rabbit Polyclonal Antibody

Supplier: Genetex

GTX107234
89319-804EA 394.91 USD
89319-804
Anti-NOTUM Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to L3MBTL2 (l(3)mbt-like 2 (Drosophila))

Suggested starting dilutions are as follows: IP: 1:500-1:1000, ChIP. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NOTUM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR