Anti-CKIP1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX107157
89319-790EA 394.79 USD
89319-790
Anti-CKIP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to NSUN6 (NOL1/NOP2/Sun domain family, member 6)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CKIP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR