Anti-FRMPD2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX107142
89319-788EA 394.91 USD
89319-788
Anti-FRMPD2 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to KLHDC7B (kelch domain containing 7B)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: KLHDC7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR