Anti-PPAPDC2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX107076
89309-746EA 427.1 USD
89309-746
Anti-PPAPDC2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CPNE9 (copine family member IX)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: PPAPDC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR