Anti-TMEM184A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106946
89319-758EA 582.76 USD
89319-758
Anti-TMEM184A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SCMH1 (sex comb on midleg homolog 1 (Drosophila))

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TMEM184A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR