Anti-TMEM184C Rabbit Polyclonal Antibody

Supplier: Genetex

GTX106935
89309-714EA 394.91 USD
89309-714
Anti-TMEM184C Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to AASDH (aminoadipate-semialdehyde dehydrogenase)

WB:Assay dependent

Type: Primary
Antigen: TMEM184C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR