Anti-SCRN3 Rabbit Polyclonal Antibody

GTX106871
89309-694EA 176.03 USD
89309-694
Anti-SCRN3 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to SYCE1 (synaptonemal complex central element protein 1)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: SCRN3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR