Anti-MFSD2B Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106799
89309-670EA 176.03 USD
89309-670
Anti-MFSD2B Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ZCCHC11 (zinc finger, CCHC domain containing 11)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: MFSD2B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR