Anti-SYNPO2 Rabbit Polyclonal Antibody

GTX106786
89309-660EA 180.24 USD
89309-660
Anti-SYNPO2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to RWDD2A (RWD domain containing 2A)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: SYNPO2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR