Anti-GNB3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106635
89319-696EA 582.56 USD
89319-696
Anti-GNB3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ITPase (inosine triphosphatase (nucleoside triphosphate pyrophosphatase))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ITPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR