Anti-RETN Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106630
89319-694EA 582.76 USD
89319-694
Anti-RETN Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FAM9B (family with sequence similarity 9, member B)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RETN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR