Anti-SH3GL2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX106471
89319-672EA 394.91 USD
89319-672
Anti-SH3GL2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SH3GL2 (SH3-domain GRB2-like 2)

Suggested starting dilutions are as follows: WB: 1:5000-1:20000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SH3GL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR