Anti-APC4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106442
89316-470EA 727.73 USD
89316-470
Anti-APC4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MID2 (midline 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: APC4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR