Anti-AK5 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX10630
89317-222EA 395.19 USD
89317-222
Anti-AK5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to STAT4

IP: Use at a concentration of 5 ?g/ml. WB: Use at a concentration of 2-3 ?g/ml. Detects a band of approximately 89 kDa (predicted molecular weight: 86 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: AK5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR