Anti-AKR1A1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX106291
89319-594EA 582.76 USD
89319-594
Anti-AKR1A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PIST (golgi associated PDZ and coiled-coil motif containing)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: AKR1A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR