Anti-AKT3 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX106259
89319-582EA 394.91 USD
89319-582
Anti-AKT3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to EB1 (microtubule-associated protein, RP/EB family, member 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: AKT3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR