Anti-ALB Rabbit Polyclonal Antibody

Supplier: Genetex

GTX106239
89319-578EA 394.91 USD
89319-578
Anti-ALB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CD158e (killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ALB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR